missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-mercaptobenzothiazole, 98%, Thermo Scientific Chemicals

Product Code. 11402768
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5331-91-9
Molecular Formula C7H4ClNS2
Molecular Weight (g/mol) 201.69
MDL Number MFCD00005783
InChI Key NKYDKCVZNMNZCM-UHFFFAOYSA-N
Synonym 5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro
PubChem CID 2723842
IUPAC Name 5-chloro-3H-1,3-benzothiazole-2-thione
SMILES ClC1=CC=C2SC(=S)NC2=C1

Specifications

Melting Point 198°C to 200°C
Quantity 25 g
Formula Weight 201.7
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-mercaptobenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'