missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyaniline hydrochloride, 98%, Thermo Scientific™

Product Code. 11435333
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4274-03-7
MDL Number MFCD00035488

Specifications

Melting Point ∼228°C (decomposition)
Molecular Formula C7H9Cl2NO
Quantity 100 g
UN Number UN2811
Synonym 5-chloro-2-methoxyaniline hydrochloride, hindamine red r, azobase cas, hindasol red rc, stabamine red rc, devol red j, fast red r base, red base ciba i, red base irga i, red salt ciba i
InChI Key ASEYMHNCCXSHIL-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)Cl)N.Cl
IUPAC Name 5-chloro-2-methoxyaniline;hydrochloride
Molecular Weight (g/mol) 194.06
PubChem CID 77957
Formula Weight 194.06
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxyaniline hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'