missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-chloro-2-methylindole, 95%, ACROS Organics™
Click to view available options
1g; Glass bottle
Specifications
CAS | 1075-35-0 |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 112°C to 118°C |
Color | Beige-Brown to Pink |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Molecular Formula | C9H8ClN |
MDL Number | MFCD00005619 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'