missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-(methylthio)aniline, 95%, Thermo Scientific™

Catalog Number 10456852
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16423-54-4
C7H8ClNS
173.658
MFCD00052816
RWUQNIPYCAHKMO-UHFFFAOYSA-N
2794577
5-chloro-2-methylsulfanylaniline
CSC1=C(C=C(C=C1)Cl)N
This item is not returnable. View return policy
16423-54-4
C7H8ClNS
173.658
MFCD00052816
RWUQNIPYCAHKMO-UHFFFAOYSA-N
2794577
5-chloro-2-methylsulfanylaniline
CSC1=C(C=C(C=C1)Cl)N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula NH2C6H3(Cl)SCH3
Quantity 1 g
Synonym 5-chloro-2-methylthio aniline, 5-chloro-2-methylsulfanyl aniline, 5-chloro-2-methylthiophenylamine, 5-chloro-2-methylthio benzenamine, 5-chloro-2-methylsulfanyl-phenylamine, benzenamine,5-chloro-2-methylthio, benzenamine, 5-chloro-2-methylthio
Formula Weight 173.67
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-(methylthio)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'