Learn More
5-chloro-2-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
£19.20 - £185.09
Chemical Identifiers
CAS | 2516-95-2 |
---|---|
Molecular Formula | C7H4ClNO4 |
Molecular Weight (g/mol) | 201.57 |
MDL Number | MFCD00007290 |
InChI Key | ZKUYSJHXBFFGPU-UHFFFAOYSA-N |
Synonym | 5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate |
PubChem CID | 17286 |
SMILES | C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10365980
|
Thermo Scientific
109680100 |
10 g | Glass bottle | N/A | |||||
10152520
|
Thermo Scientific Acros
109681000 |
100 g | Plastic Bottle | N/A | |||||
10137523
|
Thermo Scientific Acros
109685000 |
500 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2516-95-2 | |
201.57 | |
ZKUYSJHXBFFGPU-UHFFFAOYSA-N | |
17286 |
C7H4ClNO4 | |
MFCD00007290 | |
5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate | |
C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-] |
Specifications
2516-95-2 | |
100.0 | |
Green-Yellow to Yellow | |
0.5% max. | |
98.5% min. (HPLC) | |
ClC6H3(NO2)CO2H | |
09, 401 | |
Solubility in water: 9,67g/L (25°C). Other solubilities: soluble in methanol | |
C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-] | |
17286 | |
99% | |
5-Chloro-2-nitrobenzoic acid |
98.5 | |
136°C to 139°C | |
>100°C | |
Authentic | |
C7H4ClNO4 | |
MFCD00007290 | |
5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate | |
ZKUYSJHXBFFGPU-UHFFFAOYSA-N | |
201.57 | |
201.57 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 219-738-1
RUO – Research Use Only