missing translation for 'onlineSavingsMsg'
Learn More

5-chlorobenzo[d]isoxazol-3-ol, Thermo Scientific™

Product Code. 10784292
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10784292

Brand: Thermo Scientific Maybridge RF01279DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24603-63-2
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.564
InChI Key WIZWHBRFTCYPDN-UHFFFAOYSA-N
Synonym 5-chlorobenzo d isoxazol-3-ol, 5-chloro-3-hydroxybenzisoxazole, 5-chloro-1,2-benzoxazol-3-ol, 5-chloro-1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3 2h-one, 5-chloro, 5-chloro-2h-1,2-benzoxazol-3-one, pubchem24257, 5-chloro-benzo d isoxazol-3-ol, 5-chloro-benzo d isoxazol-3-one, 5-chlorobenzo d isoxazol-3 2h-one
PubChem CID 3714169
IUPAC Name 5-chloro-1,2-benzoxazol-3-one
SMILES C1=CC2=C(C=C1Cl)C(=O)NO2

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 169.57
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzo[d]isoxazol-3-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.