missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-3-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10516724
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 63417-81-2
Molecular Formula C7H5ClN2OS
Molecular Weight (g/mol) 200.64
MDL Number MFCD03407322
InChI Key YOUDLOUFERNGRO-UHFFFAOYSA-N
Synonym 5-chloromethyl-3-2-thienyl-1,2,4-oxadiazole, 5-chloromethyl-3-thiophen-2-yl-1,2,4-oxadiazole, 1,2,4-oxadiazole,5-chloromethyl-3-2-thienyl, 5-chloromethyl-3-thien-2-yl-1,2,4-oxadiazole, 5-chloromethyl-3-thiophen-2-yl-1,2,4 oxadiazole, 2-5-chloromethyl-1,2,4-oxadiazol-3-yl thiophene, 5-chloromethyl-3-thiophen-2-yl-1,2,4 oxadia zole
PubChem CID 736830
IUPAC Name 5-(chloromethyl)-3-thiophen-2-yl-1,2,4-oxadiazole
SMILES ClCC1=NC(=NO1)C1=CC=CS1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 200.65
Percent Purity 95%
Chemical Name or Material 5-(Chloromethyl)-3-(2-thienyl)-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'