missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole, 97%, Thermo Scientific™
£72.55 - £160.00
Chemical Identifiers
CAS | 1822-94-2 |
---|---|
Molecular Formula | C9H7ClN2O |
Molecular Weight (g/mol) | 194.618 |
InChI Key | XMSYUUPMPGDIOE-UHFFFAOYSA-N |
PubChem CID | 966714 |
IUPAC Name | 5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole |
SMILES | C1=CC=C(C=C1)C2=NOC(=N2)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10216422
|
Thermo Scientific Maybridge
RF01832DA |
1 g | Amber Glass Bottle |
£72.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10152844
|
Thermo Scientific Maybridge
RF01832EA |
10 g | Amber Glass Bottle |
£160.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1822-94-2 | |
194.618 | |
966714 | |
C1=CC=C(C=C1)C2=NOC(=N2)CCl |
C9H7ClN2O | |
XMSYUUPMPGDIOE-UHFFFAOYSA-N | |
5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole |
Specifications
1822-94-2 | |
C9H7ClN2O | |
XMSYUUPMPGDIOE-UHFFFAOYSA-N | |
5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole | |
966714 | |
97% |
97% | |
5-chloromethyl-3-phenyl-1,2,4-oxadiazole, 5-chloromethyl-3-phenyl-1,2,4 oxadiazole, 3-phenyl-5-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-phenyl, pubchem16349, aronis24545, 5-chloromethyl-3-phenyl-1,2,4-oxadiazol, 1,2,4-oxadiazole,5-chloromethyl-3-phenyl | |
C1=CC=C(C=C1)C2=NOC(=N2)CCl | |
194.618 | |
194.62 | |
5-(Chloromethyl)-3-phenyl-1, 2, 4-oxadiazole |