missing translation for 'onlineSavingsMsg'
Learn More
5-Chlorothiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 15408028

5-Chlorothiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 15408028
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15408028

Brand: Thermo Scientific Alfa Aesar H61474.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed container. Store away form water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42518-98-9
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00043886
InChI Key BMPDCQVRKDNUAP-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-carbonylchloride, 5-chorothiophene-2-carbonyl chloride, 5-chloro-2-thiophenecarbonyl chloride, 5-chloro-thiophene-2-carbonyl chloride, 5-chloro-2-thenoyl chloride, 2-thiophenecarbonyl chloride, 5-chloro, pubchem16132, ksc235i7t, 5-chlorothiophenecarbonyl chloride, 5-chloro-2-thenoylchloride
PubChem CID 589223
IUPAC Name 5-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 4°C
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carbonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81500169 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61474.14,H61474.06
Lot NumberP20F043 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61474.14,H61474.06
Lot Number81300282 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61474.14,H61474.06
Lot NumberU16E030 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61474.14,H61474.06
Lot NumberP04F040 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61474.14,H61474.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chlorothiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.