missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovanillic acid, tech. 90%, Thermo Scientific Chemicals

Product Code. 11405243
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62936-23-6
Molecular Formula C8H7ClO4
Molecular Weight (g/mol) 202.59
MDL Number MFCD00016531
InChI Key XBRYEHVBBMSSCG-UHFFFAOYSA-N
PubChem CID 44215
IUPAC Name 3-chloro-4-hydroxy-5-methoxybenzoic acid
SMILES COC1=C(C(=CC(=C1)C(=O)O)Cl)O

Specifications

Melting Point 242°C to 248°C
Quantity 25 g
Beilstein 2648897
Synonym 5-chlorovanillic acid, 3-chloro-4-hydroxy-5-methoxy-benzoic acid, benzoic acid, 3-chloro-4-hydroxy-5-methoxy, 3-chloro-4-hydroxy-5-methoxy benzoic acid, 5-chlorovanillicacid, 5-chloro-vanillic acid, acmc-1b2hg, chloro-4-hydroxy-3-methoxybenzoic acid, 5-chloro-4-hydroxy-3-methoxybenzoic acid, benzoic acid,3-chloro-4-hydroxy-5-methoxy
Formula Weight 202.6
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-Chlorovanillic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'