missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyano-2-fluorophenylboronic acid, 97%, Thermo Scientific™
£188.19 - £459.10
Chemical Identifiers
CAS | 468718-30-1 |
---|---|
Molecular Formula | C7H5BFNO2 |
Molecular Weight (g/mol) | 164.93 |
MDL Number | MFCD03095147 |
InChI Key | ITCSMTHUTFTXLE-UHFFFAOYSA-N |
Synonym | 5-cyano-2-fluorobenzeneboronic acid, 5-cyano-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzonitrile, boronic acid, 5-cyano-2-fluorophenyl, 5-cyano-2-fluoro-phenyl boronic acid, boronic acid, b-5-cyano-2-fluorophenyl, 5-cyano-2-fluorobenzeneboronicacid, pubchem1807, pubchem7788, acmc-1apk5 |
PubChem CID | 2783361 |
IUPAC Name | (5-cyano-2-fluorophenyl)boronic acid |
SMILES | B(C1=C(C=CC(=C1)C#N)F)(O)O |
Description
Chemical Identifiers
468718-30-1 | |
164.93 | |
ITCSMTHUTFTXLE-UHFFFAOYSA-N | |
2783361 | |
B(C1=C(C=CC(=C1)C#N)F)(O)O |
C7H5BFNO2 | |
MFCD03095147 | |
5-cyano-2-fluorobenzeneboronic acid, 5-cyano-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzonitrile, boronic acid, 5-cyano-2-fluorophenyl, 5-cyano-2-fluoro-phenyl boronic acid, boronic acid, b-5-cyano-2-fluorophenyl, 5-cyano-2-fluorobenzeneboronicacid, pubchem1807, pubchem7788, acmc-1apk5 | |
(5-cyano-2-fluorophenyl)boronic acid |
Specifications
468718-30-1 | |
100.0 | |
White | |
96% min. (HPLC) | |
C7H5BFNO2 | |
5-cyano-2-fluorobenzeneboronic acid, 5-cyano-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzonitrile, boronic acid, 5-cyano-2-fluorophenyl, 5-cyano-2-fluoro-phenyl boronic acid, boronic acid, b-5-cyano-2-fluorophenyl, 5-cyano-2-fluorobenzeneboronicacid, pubchem1807, pubchem7788, acmc-1apk5 | |
B(C1=C(C=CC(=C1)C#N)F)(O)O | |
164.93 | |
164.93 | |
Crystalline Powder |
96.0 | |
350.0°C to 353.0°C | |
Authentic | |
Glass Bottle | |
MFCD03095147 | |
ITCSMTHUTFTXLE-UHFFFAOYSA-N | |
(5-cyano-2-fluorophenyl)boronic acid | |
2783361 | |
97% | |
5-Cyano-2-fluorophenylboronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse ca
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Cyano-2-fluorophenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Cyano-2-fluorophenylboronic acid, 97%, Thermo Scientific™