missing translation for 'onlineSavingsMsg'
Learn More
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™ Product Code.: 15532313

5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™

Product Code. 15532313
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15532313

Brand: Thermo Scientific Acros 450030010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 868696-42-8
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
InChI Key OHSZRALIDGBNES-UHFFFAOYSA-N
Synonym 1,2,4-oxadiazol-3-amine, 5-cyclopropyl
PubChem CID 51072255
IUPAC Name 5-cyclopropyl-1,2,4-oxadiazol-3-amine
SMILES C1CC1C2=NC(=NO2)N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
compliance-icons
Product Identifier
  • 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.