missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 5' Fluorescent Labeled Oligo

Product Code. 11539446
Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
1 set
300pmol
Each
This item is not returnable. View return policy

Product Code. 11539446

Brand: Applied Biosystems™ 450017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5' Fluorescent Labeled Oligo

Custom 5'-labeled primers are ideal for applications including fragment analysis, gene expression studies, genotyping, and PCR. Choose among FAM™, TET™, VIC™, HEX™, NED™, and PET™ as fluorescent 5' labels. Oligos are desalted; HPLC purification is also available.
TRUSTED_SUSTAINABILITY

Specifications

Description Custom 5' Labeled Primers, 300,000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, 12 Months Shelf Life, Desalted, Option of HPLC Purification with Larger Scale, Storage at –20°C, For Use in Fragment
Product Type Custom Prism Primer
Length (Metric) 1.524 m
Content And Storage Room Temperature
Label or Dye FAM, HEX, NED, PET, TET, VIC
Primer 5' Labeled
Quantity 300,000 pmol
Grade Desalted, HPLC
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Form Solution or Dried down

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.