missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 5' Fluorescent Labeled Oligo
GREENER_CHOICE

5' Fluorescent Labeled Oligo

Brand:  Applied Biosystems™ 450007

 View more versions of this product

Product Code. 11563827

  • £111.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping, and PCR
  • All oligos are desalted; larger scales of labeled primers have an option for HPLC purification
  • Note: HPLC purification reduces the yield in half

Order Info

Shipping Condition: Dry ice

Recommended Storage

Store at -20°C. Shelf Life: 12 months from manufacturing date.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Oligonucleotides, Primers
Desalted, Option of HPLC Purification with Larger Scale
5' Labeled
HPLC
Store in Freezer (-5°C to -30°C).
6-FAM™, TET™, VIC™, HEX™, NED™, PET™
10,000 pmol
Solution or Dried down
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.