missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 5' Fluorescent Labeled Oligo

Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
1 set
300pmol
Each
Description
Custom 5'-labeled primers are ideal for applications including fragment analysis, gene expression studies, genotyping, and PCR. Choose among FAM™, TET™, VIC™, HEX™, NED™, and PET™ as fluorescent 5' labels. Oligos are desalted; HPLC purification is also available.
Specifications
Specifications
| Description | Custom 5' Fluorescent Labeled Oligo Primers, 80000 pmol, 40000 pmol Minimum HPLC Yield, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, 12 Months Shelf Life, –20°C Storage Temperature, Desalted Purification, Solution or Dried Down Format, Delivered Either in Solution or Dried Down |
| Product Type | Custom 5' Fluorescent Labeled Oligo Primers |
| Storage Buffer | TE or Water |
| Length (Metric) | 1.524 m |
| Content And Storage | Store in Freezer (-5°C to -30°C). |
| Label or Dye | FAM, HEX, NED, PET, TET, VIC |
| Primer | 5' Labeled |
| Quantity | 80,000 pmol |
| Grade | Desalted, HPLC |
| For Use With (Application) | Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction