missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 5' Fluorescent Labeled Oligo
GREENER_CHOICE

Product Code. p-7075580
Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
1 set
300pmol
Each
This item is not returnable. View return policy

Product Code. 11563827

Brand: Applied Biosystems™ 450007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5' Fluorescent Labeled Oligo

  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping, and PCR
  • All oligos are desalted; larger scales of labeled primers have an option for HPLC purification
  • Note: HPLC purification reduces the yield in half

Order Info

Shipping Condition: Dry ice

Recommended Storage

Store at -20°C. Shelf Life: 12 months from manufacturing date.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Oligonucleotides, Primers
Content And Storage Store in Freezer (-5°C to -30°C).
Purification Method Desalted, Option of HPLC Purification with Larger Scale
Label or Dye 6-FAM™, TET™, VIC™, HEX™, NED™, PET™
Primer 5' Labeled
Quantity 10,000 pmol
Grade HPLC
Form Solution or Dried down

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.