missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-methoxybenzeneboronic acid, 98%, Thermo Scientific™
£46.10
Chemical Identifiers
CAS | 179897-94-0 |
---|---|
Molecular Formula | C7H8BFO3 |
Molecular Weight (g/mol) | 169.95 |
MDL Number | MFCD01863526 |
InChI Key | CCQKIRUMTHHPSX-UHFFFAOYSA-N |
Synonym | 5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl |
PubChem CID | 2782673 |
IUPAC Name | (5-fluoro-2-methoxyphenyl)boronic acid |
SMILES | COC1=CC=C(F)C=C1B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465507
|
Thermo Scientific Alfa Aesar
H32748.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
179897-94-0 | |
169.95 | |
CCQKIRUMTHHPSX-UHFFFAOYSA-N | |
2782673 | |
COC1=CC=C(F)C=C1B(O)O |
C7H8BFO3 | |
MFCD01863526 | |
5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl | |
(5-fluoro-2-methoxyphenyl)boronic acid |
Specifications
179897-94-0 | |
MFCD01863526 | |
CCQKIRUMTHHPSX-UHFFFAOYSA-N | |
(5-fluoro-2-methoxyphenyl)boronic acid | |
2782673 | |
98% |
C7H8BFO3 | |
5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl | |
COC1=CC=C(F)C=C1B(O)O | |
169.95 | |
169.95 | |
5-Fluoro-2-methoxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures