Learn More
5-Fluoro-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
£23.00 - £44.90
Chemical Identifiers
CAS | 445-05-6 |
---|---|
Molecular Formula | C7H6ClFO2S |
Molecular Weight (g/mol) | 208.63 |
MDL Number | MFCD00052958 |
InChI Key | NPEWBMJAUUSBLE-UHFFFAOYSA-N |
Synonym | 5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride |
PubChem CID | 136286 |
SMILES | CC1=CC=C(F)C=C1S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10755933
|
Thermo Scientific Maybridge
TL00317EA |
10 g |
£23.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10226004
|
Thermo Scientific Maybridge
TL00317EB |
25 g |
£44.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
445-05-6 | |
208.63 | |
NPEWBMJAUUSBLE-UHFFFAOYSA-N | |
136286 |
C7H6ClFO2S | |
MFCD00052958 | |
5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride | |
CC1=CC=C(F)C=C1S(Cl)(=O)=O |
Specifications
445-05-6 | |
97% | |
C7H6ClFO2S | |
MFCD00052958 | |
NPEWBMJAUUSBLE-UHFFFAOYSA-N | |
5-fluoro-2-methylbenzene-1-sulfonyl chloride | |
136286 | |
97% |
97.0 | |
Amber glass bottle | |
F(CH3)C6H3SO2Cl | |
5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride | |
CC1=CC=C(F)C=C1S(Cl)(=O)=O | |
208.63 | |
208.64 | |
5-Fluoro-2-methylbenzene-1-sulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.