missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorocytosine, 99+%, Thermo Scientific Chemicals

£65.12 - £463.31

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
IUPAC Name 6-amino-5-fluoro-1H-pyrimidin-2-one
SMILES NC1=C(F)C=NC(=O)N1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10125793
View Documents Promotion Details
Thermo Scientific Acros
258342500
250 mg Glass bottle
£65.12
250mg
Please to purchase this item. Need a web account? Register with us today!
10204381
View Documents
Thermo Scientific Acros
258340010
1 g Glass bottle
£154.77
1g
Please to purchase this item. Need a web account? Register with us today!
10742354
View Documents
Thermo Scientific Acros
258340050
5 g Glass bottle
£463.31
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2022-85-7
129.09
XRECTZIEBJDKEO-UHFFFAOYSA-N
3366
6-amino-5-fluoro-1H-pyrimidin-2-one
C4H4FN3O
MFCD00006035,MFCD00179326,MFCD03547958
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
CHEBI:5100
NC1=C(F)C=NC(=O)N1

Specifications

2022-85-7
100.0
White
Authentic
C4H4FN3O
25, 3688
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
XRECTZIEBJDKEO-UHFFFAOYSA-N
6-amino-5-fluoro-1H-pyrimidin-2-one
3366
129.09
Crystalline Powder
99.0
296.0°C
1.0% max. (1 g, 105°C)
99+%
MFCD00006035,MFCD00179326,MFCD03547958
15, 4156
Solubility in water: 15g/L (25°C)
NC1=C(F)C=NC(=O)N1
129.09
CHEBI:5100
99+%
5-Fluorocytosine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 217-968-7

Product Identifier
  • 5-Fluorocytosine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.