missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorouracil, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Anticancer agent. Metabolized to form fluorodeoxyuridine monophosphate (FdUMP), fluorodeoxyuridine triphosphate (FdUTP) and fluorouridine (FUTP). FdUMP inhibits thymidylate synthase, causing a reduction in dTMP synthesis. FUTP and FdUTP are misincorporated into RNA and DNA respectively.
Specifications
Specifications
CAS | 51-21-8 |
Molecular Formula | C4H3FN2O2 |
MDL Number | MFCD00006018 |
Quantity | 50 mg |
Synonym | 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton |
InChI Key | GHASVSINZRGABV-UHFFFAOYSA-N |
SMILES | FC1=CNC(=O)NC1=O |
IUPAC Name | 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione |
Molecular Weight (g/mol) | 130.08 |
PubChem CID | 3385 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'