missing translation for 'onlineSavingsMsg'
Learn More

5-Hex-1-ynylnicotinic acid, 97%, Thermo Scientific™

Product Code. 10161312
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10161312

Brand: Thermo Scientific Maybridge KM09434DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-31-9
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00830967
InChI Key FKWDNFDBDZUPIQ-UHFFFAOYSA-N
Synonym 5-hex-1-ynylnicotinic acid, 5-hex-1-ynylnicotinc acid, 3-pyridinecarboxylicacid, 5-1-hexyn-1-yl, 5-hex-1-yn-1-yl pyridine-3-carboxylic acid, 5-hex-1-ynyl-nicotinic acid, 5-hex-1-ynyl nicotinic acid, 5-hex-1-yn-1-yl nicotinic acid
PubChem CID 2736516
IUPAC Name 5-hex-1-ynylpyridine-3-carboxylic acid
SMILES CCCCC#CC1=CC(=CN=C1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 5-Hex-1-ynylnicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.