missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenoic acid, 98%, Thermo Scientific Chemicals

Product Code. 13448668
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1577-22-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
InChI Key XUDOZULIAWNMIU-UHFFFAOYSA-N
Synonym 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
PubChem CID 15308
IUPAC Name hex-5-enoic acid
SMILES C=CCCCC(=O)O

Specifications

Density 0.9610g/mL
Boiling Point 203.0°C
Flash Point 104°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2=CH(CH2)3CO2H
Quantity 1 g
Specific Gravity 0.961
Solubility Information Solubility in water: insoluble.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material 5-Hexenoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hexenoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.