missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexenoic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
5-Hexenoic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hexenoic acid is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
5-Hexenoic acid is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.961 |
Boiling Point | 107°C (17 mmHg) |
Flash Point | 104°C (219°F) |
Refractive Index | 1.434 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 1743172 |
Solubility Information | Insoluble in water. |
Formula Weight | 114.14 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'