missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynenitrile, 98%, Thermo Scientific Chemicals

Catalog Number 11397755
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14918-21-9
C6H7N
93.129
MFCD00001978
JZYKFLLRVPPISG-UHFFFAOYSA-N
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g
139852
hex-5-ynenitrile
C#CCCCC#N
This item is not returnable. View return policy
14918-21-9
C6H7N
93.129
MFCD00001978
JZYKFLLRVPPISG-UHFFFAOYSA-N
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g
139852
hex-5-ynenitrile
C#CCCCC#N

5-Hexynenitrile is used in the preparation of annelated symmetric and asymmetric 3,3'-disubstituted 2,2'-bipyridines by cobalt(I)-catalyzed [2+2+2] cycloadditions with 1,3-diynes. Further, it reacts with 3-bromo-pyridine to prepare 6-(3-pyridinyl)-5-hexynenitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexynenitrile is used in the preparation of annelated symmetric and asymmetric 3,3′-disubstituted 2,2′-bipyridines by cobalt(I)-catalyzed [2+2+2] cycloadditions with 1,3-diynes. Further, it reacts with 3-bromo-pyridine to prepare 6-(3-pyridinyl)-5-hexynenitrile.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 0.889
Boiling Point 115°C to 117°C
Flash Point 42°C (107°F)
Refractive Index 1.441
Quantity 1 g
UN Number UN1992
Beilstein 1735926
Solubility Information Miscible with water.
Formula Weight 93.13
Percent Purity 98%
Chemical Name or Material 5-Hexynenitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'