missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-2-iodobenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 248.01 |
Percent Purity | 95% |
Chemical Name or Material | 5-Hydroxy-2-iodobenzaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction