missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-2-nitrobenzoic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 5-hydroxy-2-nitrobenzoic acid, 3-carboxy-4-nitrophenol, benzoic acid, 5-hydroxy-2-nitro, 5-hydroxy-2-nitro-benzoic acid, 2-nitro-5-oxybenzoic acid, acmc-1b06l, 4-10-00-00338 beilstein handbook reference, 2-nitro-5-hydroxybenzoic acid, benzoic acid,5-hydroxy-2-nitro, 3-carboxy-4-nitrophenol;2-nitro-5-hydroxybenzoic acid |
Formula Weight | 183.12 |
Percent Purity | 95% |
Chemical Name or Material | 5-Hydroxy-2-nitrobenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'