missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)furfural, 98%, Thermo Scientific Chemicals

£41.91 - £644.21

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10654972
View Documents Promotion Details
Thermo Scientific Acros
121460010
1 g Glass bottle
£41.91
1g
Please to purchase this item. Need a web account? Register with us today!
10316520
View Documents Promotion Details
Thermo Scientific Acros
121460050
5 g Glass bottle
£165.83
5g
Please to purchase this item. Need a web account? Register with us today!
11986161
View Documents
Thermo Scientific Acros
121460250
25 g Glass bottle
£644.21
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-47-0
126.11
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
CHEBI:412516
C1=C(OC(=C1)C=O)CO
C6H6O3
NOEGNKMFWQHSLB-UHFFFAOYSA-N
237332
5-(hydroxymethyl)furan-2-carbaldehyde

Specifications

67-47-0
Yellow
79°C
97.5% min. (GC)
1.5627
15,4871
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
C1=C(OC(=C1)C=O)CO
126.11
CHEBI:412516
98%
5-(Hydroxymethyl)furfural
30.0°C to 34.0°C
114°C to 116°C (1.0 mmHg)
Authentic
C6H6O3
18, V,1, 130
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
NOEGNKMFWQHSLB-UHFFFAOYSA-N
5-(hydroxymethyl)furan-2-carbaldehyde
237332
126.11
Crystalline Powder, Chunks or Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-654-9

Product Identifier
  • 5-(Hydroxymethyl)furfural

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only