missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hydroxymethylcytosine Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540097
Code : NEW
Additional Details : Weight : 0.00900kg
Description
5-hydroxymethylcytosine (5-hmC) has been recently discovered in mammalian DNA. 5-hmC results from the enzymatic conversion of 5-methylcytosine into 5-hydroxymethylcytosine by the TET family of oxygenases. Initially, the 5-hmC base had been identified in Purkinje neurons, in granule cells and embryonic stem cells where it is present at high levels (up to 0,6% of total nucleotides in Purkinje cells). A recent report indicates that 5-hmC is also abundant in brain tissue, especially in areas that are associated with higher cognitive functions.Early evidence suggests that 5-hmC may represent an intermediate in a new pathway which demethylates DNA, converting 5-mC to cytosine. Although its precise role has still to be shown, recent results indicate, however, that 5-hmC may play important roles distinct from 5-mC. This may open up entirely new perspectives in epigenetic studies.Specifications
5-hydroxymethylcytosine | |
Polyclonal | |
Unconjugated | |
5hmc; 5-hmc; 5-HydroxymethylCytidine; C5H7N3O2 | |
Human KLH-conjugated 5-hydroxymethylcytosine. | |
50 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunoprecipitation, Peptide Array | |
3.5 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Human, Mouse | |
Antibody | |
IgG |