Learn More
Molecular Probes™ 5-IAF (5-Iodoacetamidofluorescein)
5-IAF (5-Iodoacetamidofluorescein)
Brand: Molecular Probes™ I30451
Code : 50
Additional Details : Weight : 0.00525kg
Description
The thiol-reactive 5-iodoacetamidofluorescein (5-IAF) can be used to produce bioconjugates with the 5-isomer of fluorescein.Specifications
25 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
FITC (Fluorescein) |
5-IAF | |
Thiol | |
Alkyl Halide, Iodoacetamide | |
Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.