missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10452684
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 34091-51-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD11109327
InChI Key RJYWUQWCLZYCTI-UHFFFAOYSA-N
PubChem CID 13334065
SMILES CN1N=CC=C1I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-iodo-1-methyl-1h-pyrazole, 1-methyl-5-iodopyrazole, 1h-pyrazole,5-iodo-1-methyl, 1h-pyrazole, 5-iodo-1-methyl, 1-methyl-5-iodo-1h-pyrazole
IUPAC Name 5-iodo-1-methyl-1H-pyrazole
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 5-Iodo-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'