missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-phenoxypyridine, ≥95%, Thermo Scientific™

£55.20 - £138.00

Chemical Identifiers

CAS 352018-92-9
Molecular Formula C11H8INO
Molecular Weight (g/mol) 297.095
MDL Number MFCD02681952
InChI Key JCEXQQNAXWYXRX-UHFFFAOYSA-N
Synonym 5-iodo-2-phenoxy-pyridine, pyridine,5-iodo-2-phenoxy
PubChem CID 2776503
IUPAC Name 5-iodo-2-phenoxypyridine
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10304282
View Documents Promotion Details
Thermo Scientific Maybridge
CC19611DA
1 g
£55.20
1g
Please to purchase this item. Need a web account? Register with us today!
10503002
View Documents Promotion Details
Thermo Scientific Maybridge
CC19611EA
10 g
£138.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

352018-92-9
297.095
JCEXQQNAXWYXRX-UHFFFAOYSA-N
2776503
C1=CC=C(C=C1)OC2=NC=C(C=C2)I
C11H8INO
MFCD02681952
5-iodo-2-phenoxy-pyridine, pyridine,5-iodo-2-phenoxy
5-iodo-2-phenoxypyridine

Specifications

352018-92-9
100.0
Amber Glass Bottle
MFCD02681952
JCEXQQNAXWYXRX-UHFFFAOYSA-N
5-iodo-2-phenoxypyridine
2776503
95%
94.5
95%
C11H8INO
5-iodo-2-phenoxy-pyridine, pyridine,5-iodo-2-phenoxy
C1=CC=C(C=C1)OC2=NC=C(C=C2)I
297.095
297.09
5-Iodo-2-phenoxypyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications