missing translation for 'onlineSavingsMsg'
Learn More
5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™ Product Code.: 10522334

5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Product Code. 10522334
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10522334

Brand: Thermo Scientific Maybridge CC41406CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-16-8
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD09702358
InChI Key RDQRMCJCROUHOF-UHFFFAOYSA-N
PubChem CID 24229489
SMILES CN1C=CC2=CC(=CC=C12)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-isocyanato-1-methyl-1h-indole, 1h-indole, 5-isocyanato-1-methyl, 1-methylindol-5-isocyanate, 1h-indole,5-isocyanato-1-methyl, 1-methyl-1h-indole-5-yl isocyanate
IUPAC Name 5-isocyanato-1-methylindole
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 5-Isocyanato-1-methyl-1H-indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.