missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxy-2-methyl-3-indoleacetic acid, 98%, Thermo Scientific Chemicals
£70.00 - £223.00
Chemical Identifiers
CAS | 2882-15-7 |
---|---|
Molecular Formula | C12H13NO3 |
Molecular Weight (g/mol) | 219.24 |
MDL Number | MFCD00005618 |
InChI Key | TXWGINUZLBAKDF-UHFFFAOYSA-N |
Synonym | 5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid |
PubChem CID | 76151 |
IUPAC Name | 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid |
SMILES | CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10183620
|
Thermo Scientific Acros
147510010 |
1 g | Glass bottle |
£70.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10367120
|
Thermo Scientific Acros
147510050 |
5 g | Glass bottle |
£223.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2882-15-7 | |
219.24 | |
TXWGINUZLBAKDF-UHFFFAOYSA-N | |
76151 | |
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O |
C12H13NO3 | |
MFCD00005618 | |
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid | |
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid |
Specifications
2882-15-7 | |
100.0 | |
Beige or White to Yellow | |
98% | |
MFCD00005618 | |
Solubility in water: insoluble in water | |
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O | |
219.24 | |
219.24 | |
Crystalline Powder |
97.5 | |
162°C to 165°C | |
Authentic | |
C12H13NO3 | |
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid | |
TXWGINUZLBAKDF-UHFFFAOYSA-N | |
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid | |
76151 | |
98% | |
5-Methoxy-2-methyl-3-indoleacetic acid, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 220-734-7
RUO – Research Use Only