Learn More
5-Methoxyisatin, 97%, Thermo Scientific Chemicals
£38.30 - £566.00
Chemical Identifiers
CAS | 39755-95-8 |
---|---|
Molecular Formula | C9H7NO3 |
Molecular Weight (g/mol) | 177.16 |
MDL Number | MFCD00169023 |
InChI Key | DMHGXMPXHPOXBF-UHFFFAOYSA-N |
Synonym | 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci |
PubChem CID | 38333 |
IUPAC Name | 5-methoxy-1H-indole-2,3-dione |
SMILES | COC1=CC=C2NC(=O)C(=O)C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13545030
|
Thermo Scientific Acros
448830050 |
5 g |
£38.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13555030
|
Thermo Scientific Acros
448830250 |
25 g |
£150.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13565030
|
Thermo Scientific Acros
448831000 |
100 g |
£566.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39755-95-8 | |
177.16 | |
DMHGXMPXHPOXBF-UHFFFAOYSA-N | |
38333 | |
COC1=CC=C2NC(=O)C(=O)C2=C1 |
C9H7NO3 | |
MFCD00169023 | |
5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci | |
5-methoxy-1H-indole-2,3-dione |
Spezifikation
39755-95-8 | |
96% min. (GC) | |
C9H7NO3 | |
5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci | |
COC1=CC=C2NC(=O)C(=O)C2=C1 | |
177.16 | |
177.16 | |
5-Methoxyisatin |
201°C to 203°C | |
Glass bottle | |
MFCD00169023 | |
DMHGXMPXHPOXBF-UHFFFAOYSA-N | |
5-methoxy-1H-indole-2,3-dione | |
38333 | |
97% |
Sicherheit und Handhabung
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
- 5-Methoxyisatin
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only