missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11399615
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-83-1
Molecular Formula C11H15ClN2O
Molecular Weight (g/mol) 226.704
MDL Number MFCD00012684
InChI Key TXVAYRSEKRMEIF-UHFFFAOYSA-N
Synonym 5-methoxytryptamine hydrochloride, mexamine hydrochloride, o-methylserotonin hydrochloride, 2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methoxyindole hydrochloride, unii-4265ib6qek, 5-methoxy-1h-indole-3-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride, 5-methoxy tryptamine hcl, 5 methoxytryptamine hydrochloride
PubChem CID 6198
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES COC1=CC2=C(C=C1)NC=C2CCN.Cl

Specifications

Melting Point 245°C to 250°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 3717598
Sensitivity Hygroscopic
Merck Index 14,6002
Formula Weight 226.71
Percent Purity ≥98%
Chemical Name or Material 5-Methoxytryptamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'