missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate, 99%, Thermo Scientific™

Product Code. 11409290
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187931-57-2
Molecular Formula C6H9N3O5
Molecular Weight (g/mol) 203.15
MDL Number MFCD07781053
InChI Key BQNHEXDOLOGXGU-UHFFFAOYSA-N
Synonym 5-methyl-1,3,4-oxadiazol-2-yl methanamine oxalate, c-5-methyl-1,3,4 oxadiazol-2-yl-methylamine oxalate, 5-methyl-1,3,4-oxadiazole-2-methylamine oxalate, c4h7n3o.c2h2o4, c-5-methyl-1,3,4 oxadiazol-2yl-methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methanamine; oxalic acid
PubChem CID 53445292
IUPAC Name (5-methyl-1,3,4-oxadiazol-2-yl)methanamine;oxalic acid
SMILES OC(=O)C(O)=O.CC1=NN=C(CN)O1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 203.15
Percent Purity 99%
Chemical Name or Material 5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'