missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate, 99%, Thermo Scientific™
£61.80 - £217.00
Chemical Identifiers
CAS | 1187931-57-2 |
---|---|
Molecular Formula | C6H9N3O5 |
Molecular Weight (g/mol) | 203.15 |
MDL Number | MFCD07781053 |
InChI Key | BQNHEXDOLOGXGU-UHFFFAOYSA-N |
Synonym | 5-methyl-1,3,4-oxadiazol-2-yl methanamine oxalate, c-5-methyl-1,3,4 oxadiazol-2-yl-methylamine oxalate, 5-methyl-1,3,4-oxadiazole-2-methylamine oxalate, c4h7n3o.c2h2o4, c-5-methyl-1,3,4 oxadiazol-2yl-methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methanamine; oxalic acid |
PubChem CID | 53445292 |
IUPAC Name | (5-methyl-1,3,4-oxadiazol-2-yl)methanamine;oxalic acid |
SMILES | OC(=O)C(O)=O.CC1=NN=C(CN)O1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
Chemical Identifiers
1187931-57-2 | |
203.15 | |
BQNHEXDOLOGXGU-UHFFFAOYSA-N | |
53445292 | |
OC(=O)C(O)=O.CC1=NN=C(CN)O1 |
C6H9N3O5 | |
MFCD07781053 | |
5-methyl-1,3,4-oxadiazol-2-yl methanamine oxalate, c-5-methyl-1,3,4 oxadiazol-2-yl-methylamine oxalate, 5-methyl-1,3,4-oxadiazole-2-methylamine oxalate, c4h7n3o.c2h2o4, c-5-methyl-1,3,4 oxadiazol-2yl-methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methanamine; oxalic acid | |
(5-methyl-1,3,4-oxadiazol-2-yl)methanamine;oxalic acid |
Specifications
1187931-57-2 | |
MFCD07781053 | |
5-methyl-1,3,4-oxadiazol-2-yl methanamine oxalate, c-5-methyl-1,3,4 oxadiazol-2-yl-methylamine oxalate, 5-methyl-1,3,4-oxadiazole-2-methylamine oxalate, c4h7n3o.c2h2o4, c-5-methyl-1,3,4 oxadiazol-2yl-methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methylamine oxalate, 1-5-methyl-1,3,4-oxadiazol-2-yl methanamine; oxalic acid | |
OC(=O)C(O)=O.CC1=NN=C(CN)O1 | |
203.15 | |
203.15 | |
5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate |
C6H9N3O5 | |
Air Sensitive | |
BQNHEXDOLOGXGU-UHFFFAOYSA-N | |
(5-methyl-1,3,4-oxadiazol-2-yl)methanamine;oxalic acid | |
53445292 | |
99% |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen