missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10701094
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 849776-88-1
Molecular Formula C16H21BN2O2
Molecular Weight (g/mol) 284.17
MDL Number MFCD06797463
InChI Key XSUARWLKVRBBKT-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 5-methyl-1-phenyl-1h-pyrazole-4-boronic acid, pinacol ester, 5-methyl-1-phenyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, pubchem18509, 5-methyl-1-phenyl-1h-pyrazole-4-boronic acid pinacol ester, 5-methyl-1-phenylpyrazole-4-boronic acid pinacol ester, 5-methyl-1-phenyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1h-pyrazole, 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2795327
IUPAC Name 5-methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CC1=C(C=NN1C1=CC=CC=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 284.16
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.