missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2(3H)-benzoxazolethione, 97%, Thermo Scientific™

Product Code. 11438877
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22876-22-8
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00052492
InChI Key SSWZUOXLFTXIEZ-UHFFFAOYSA-N
Synonym 2-mercapto-5-methylbenzoxazole, 5-methyl-1,3-benzoxazole-2-thiol, 5-methylbenzo d oxazole-2 3h-thione, 5-methylbenzo d oxazole-2-thiol, 5-methylbenzoxazoline-2-thione, 5-methyl-1,3-benzoxazole-2 3h-thione, 5-methyl-2 3h-benzoxazolethione, 5-methyl-2-mercaptobenzoxazole, 5-methyl-2,3-dihydro-1,3-benzoxazole-2-thione, 2 3h-benzoxazolethione, 5-methyl
PubChem CID 710258
IUPAC Name 5-methyl-3H-1,3-benzoxazole-2-thione
SMILES CC1=CC2=C(C=C1)OC(=S)N2

Specifications

Melting Point 223°C to 225°C
Quantity 1 g
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-2(3H)-benzoxazolethione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'