missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11410813
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

Solubility
Soluble in alcohol and water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Density 1.106
Boiling Point 186°C to 188°C
Flash Point 72°C (161°F)
Refractive Index 1.531
Quantity 500 g
Beilstein 106895
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol and water.
Formula Weight 110.11
Percent Purity 98%
Chemical Name or Material 5-Methyl-2-furaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'