missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10100682
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 17515-74-1
Molecular Formula C7H5F3O3
Molecular Weight (g/mol) 194.11
MDL Number MFCD00275563
InChI Key NURNFMNHTHXTDS-UHFFFAOYSA-N
PubChem CID 2782183
IUPAC Name 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
SMILES CC1=CC(C(O)=O)=C(O1)C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl
Formula Weight 194.11
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.