missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 5 g |
Synonym | 5-methyl-2-trifluoromethyl furan-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furoic acid, 5-methyl-2-trifluoromethyl-furan-3-carboxylic acid, 2-trifluoromethyl-5-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-methyl-2-trifluoromethyl, 2-trifluoromethyl-5-methyl-3-furoic acid, 2-trifluoromethyl-5-methylfuran-3-carboxylic acid, 5-methyl-2-trifluoromethyl-3-furancarboxylic acid, 3-furancarboxylic acid,5-methyl-2-trifluoromethyl |
Formula Weight | 194.11 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methyl-2-(trifluoromethyl)-3-furoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction