missing translation for 'onlineSavingsMsg'
Learn More

[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine, 97%, Thermo Scientific™

Product Code. 10653013
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-05-7
Molecular Formula C7H8F3NO
Molecular Weight (g/mol) 179.14
MDL Number MFCD02180792
InChI Key ROYYYTOVBUUPDX-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl-3-furyl methylamine, 5-methyl-2-trifluoromethyl furan-3-yl methanamine, 3-aminomethyl-5-methyl-2-trifluoromethyl furan, 1-5-methyl-2-trifluoromethyl furan-3-yl methanamine, 5-methyl-2-trifluoromethyl-3-furanyl methanamine
PubChem CID 2779900
IUPAC Name [5-methyl-2-(trifluoromethyl)furan-3-yl]methanamine
SMILES CC1=CC(CN)=C(O1)C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 179.14
Percent Purity 97%
Chemical Name or Material [5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'