missing translation for 'onlineSavingsMsg'
Learn More

5-methyl-3-isoxazolecarbaldehyde, 97%, Thermo Scientific™

Product Code. 10276642
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 62254-74-4
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.10
MDL Number MFCD06797459
InChI Key OCOCVXUEFWNRJU-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carbaldehyde, 5-methylisoxazole-3-carboxaldehyde, 5-methyl-3-isoxazolecarbaldehyde, 3-isoxazolecarboxaldehyde, 5-methyl, 5-methyl-isoxazole-3-carbaldehyde, pubchem11024, acmc-1b6yu, 5-methyl-3-formyl-isoxazole, 5-methylisoxazole-3-aldehyde, 5-methylisoxazol-3-carbaldehyde
PubChem CID 2795222
IUPAC Name 5-methyl-1,2-oxazole-3-carbaldehyde
SMILES CC1=CC(C=O)=NO1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-isoxazolecarbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'