missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-methyl-3-isoxazolecarbaldehyde, 97%, Thermo Scientific™
£54.00
Chemical Identifiers
CAS | 62254-74-4 |
---|---|
Molecular Formula | C5H5NO2 |
Molecular Weight (g/mol) | 111.10 |
MDL Number | MFCD06797459 |
InChI Key | OCOCVXUEFWNRJU-UHFFFAOYSA-N |
Synonym | 5-methylisoxazole-3-carbaldehyde, 5-methylisoxazole-3-carboxaldehyde, 5-methyl-3-isoxazolecarbaldehyde, 3-isoxazolecarboxaldehyde, 5-methyl, 5-methyl-isoxazole-3-carbaldehyde, pubchem11024, acmc-1b6yu, 5-methyl-3-formyl-isoxazole, 5-methylisoxazole-3-aldehyde, 5-methylisoxazol-3-carbaldehyde |
PubChem CID | 2795222 |
IUPAC Name | 5-methyl-1,2-oxazole-3-carbaldehyde |
SMILES | CC1=CC(C=O)=NO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10276642
|
Thermo Scientific Maybridge
CC10404DA |
1 g | Amber Glass Bottle | ||||||
Description
Chemical Identifiers
62254-74-4 | |
111.10 | |
OCOCVXUEFWNRJU-UHFFFAOYSA-N | |
2795222 | |
CC1=CC(C=O)=NO1 |
C5H5NO2 | |
MFCD06797459 | |
5-methylisoxazole-3-carbaldehyde, 5-methylisoxazole-3-carboxaldehyde, 5-methyl-3-isoxazolecarbaldehyde, 3-isoxazolecarboxaldehyde, 5-methyl, 5-methyl-isoxazole-3-carbaldehyde, pubchem11024, acmc-1b6yu, 5-methyl-3-formyl-isoxazole, 5-methylisoxazole-3-aldehyde, 5-methylisoxazol-3-carbaldehyde | |
5-methyl-1,2-oxazole-3-carbaldehyde |
Specifications
62254-74-4 | |
C5H5NO2 | |
5-methylisoxazole-3-carbaldehyde, 5-methylisoxazole-3-carboxaldehyde, 5-methyl-3-isoxazolecarbaldehyde, 3-isoxazolecarboxaldehyde, 5-methyl, 5-methyl-isoxazole-3-carbaldehyde, pubchem11024, acmc-1b6yu, 5-methyl-3-formyl-isoxazole, 5-methylisoxazole-3-aldehyde, 5-methylisoxazol-3-carbaldehyde | |
CC1=CC(C=O)=NO1 | |
111.10 | |
111.1 | |
5-Methyl-3-isoxazolecarbaldehyde |
97% | |
MFCD06797459 | |
OCOCVXUEFWNRJU-UHFFFAOYSA-N | |
5-methyl-1,2-oxazole-3-carbaldehyde | |
2795222 | |
97% |