missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-isoxazolecarbonitrile, 97%, Thermo Scientific™

Product Code. 10328113
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 57351-99-2
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.10
MDL Number MFCD09064940
InChI Key JTPIMYCMWLOEDD-UHFFFAOYSA-N
PubChem CID 24229456
IUPAC Name 5-methyl-1,2-oxazole-3-carbonitrile
SMILES CC1=CC(=NO1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 5-methylisoxazole-3-carbonitrile, 5-methyl-3-isoxazolecarbonitrile, 5-methyl-isoxazole-3-carbonitrile, 3-cyano-5-methylisoxazole, 3-isoxazolecarbonitrile,5-methyl, 3-isoxazolecarbonitrile, 5-methyl, 5-?methylisoxazole-?3-?carbonitrile
Formula Weight 108.1
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-isoxazolecarbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'