missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-3-phenyl-4-isoxazolyl)methylamine, 97%, Thermo Scientific™

Product Code. 10101212
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-01-3
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD02677687
InChI Key KJGJWCJXSLAKKS-UHFFFAOYSA-N
Synonym 5-methyl-3-phenyl-4-isoxazolyl methylamine, 5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, 4-isoxazolemethanamine,5-methyl-3-phenyl, 4-aminomethyl-5-methyl-3-phenylisoxazole, 1-5-methyl-3-phenyl-1,2-oxazol-4-yl methanamine, 3-phenyl-5-methylisoxazole-4-methaneamine, 5-methyl-3-phenylisoxazol-4-yl methanamine, 5-methyl-3-phenylisoxazol-4-yl methylamine, 5-methyl-3-phenylisoxazol-4-yl methyl amine, 1-5-methyl-3-phenylisoxazol-4-yl methylamine
PubChem CID 2776151
IUPAC Name (5-methyl-3-phenyl-1,2-oxazol-4-yl)methanamine
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material (5-Methyl-3-phenyl-4-isoxazolyl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.