missing translation for 'onlineSavingsMsg'
Learn More
5-Methylcyclohexane-1,3-dione, 98%, Thermo Scientific Chemicals Product Code.: 11422348

5-Methylcyclohexane-1,3-dione, 98%, Thermo Scientific Chemicals

Product Code. 11422348
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11422348

Brand: Thermo Scientific Alfa Aesar A15280.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4341-24-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00010379
InChI Key DMIIMPQQPXUKOO-UHFFFAOYSA-N
PubChem CID 458095
IUPAC Name 5-methylcyclohexane-1,3-dione
SMILES CC1CC(=O)CC(=O)C1

Specifications

Melting Point 126°C to 130°C
Quantity 1 g
Beilstein 1099692
Synonym 5-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-methyl, 5-methyl-cyclohexane-1,3-dione, pubchem14279, acmc-1aftb, 5-methylcyclohexane dione, 1,3-dioxo-5-methylcyclohexane, 1,3-cyclohexanedione,5-methyl, 5-methyl cyclohexane-1,3-dione
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material 5-Methylcyclohexane-1,3-dione

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237817 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15280.06,A15280.03,A15280.14
Lot Number10170107 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15280.06,A15280.03,A15280.14
Lot NumberD27U008 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15280.06,A15280.03,A15280.14
Lot NumberJ9552A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15280.06,A15280.03,A15280.14
Lot Number10171261 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15280.06,A15280.03,A15280.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methylcyclohexane-1,3-dione, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.