missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, 98+%, Thermo Scientific Chemicals

Product Code. 10591981
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Acidity 10mg KOH/g max.
CAS Min % 98.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1000g/mL
Boiling Point 187°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 25 g
Beilstein 17, 289
Specific Gravity 1.1
Solubility Information Solubility in water: 33.3g/L (20 c)
Formula Weight 110.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 5-Methylfurfural, 98+%
Show More Show Less
Product Identifier
  • 5-Methylfurfural

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'