missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11479316
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-46-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00036494
InChI Key MHPUGCYGQWGLJL-UHFFFAOYSA-N
Synonym isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
PubChem CID 12344
IUPAC Name 5-methylhexanoic acid
SMILES CC(C)CCCC(O)=O

Specifications

Density 0.91
Boiling Point 216°C
Flash Point 102°C (216°F)
Odor Rancid
Refractive Index 1.422
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 5-Methylhexanoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'